Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)cobalt(III) Hexafluorophosphate 96.0+%, TCI America™
SDP

Catalog No. C15891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15891G Supplier TCI America Supplier No. C15891G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12427-42-8
Molecular Formula C10H10CoF6P
MDL Number MFCD00011874
Synonym Cobaltocene Hexafluorophosphate

Specifications

Color Yellow
Quantity 1 g
Formula Weight 334.09
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(cyclopentadienyl)cobalt(III) Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.