CAS RN 1243244-14-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Porcn Inhibitor IV, LGK974
A cell-permeable inhibitor of Wnt secretion from L-cell Wnt1/2/3/3a/6/7a/9a transfectants (IC50 from 50 pM to 1.1 nM).