Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Porcn Inhibitor IV, LGK974

Catalog No. 5310910001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-109-10001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-109-10001 Supplier MilliporeSigma™ Supplier No. 5310910001
Add to Cart
Add to Cart

A cell-permeable inhibitor of Wnt secretion from L-cell Wnt1/2/3/3a/6/7a/9a transfectants (IC50 from 50 pM to 1.1 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 97% by HPLC
Target Proteins Porcupine
CAS 1243244-14-5
Molecular Formula C23H20N6O
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.