accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNH3CCH3CH3CH3NH3CH3CH3CCH3HOOH

CAS RN 1243264-54-1

IUPAC Name: {2,6-bis[(2,2,6,6-tetramethylpiperidin-1-yl)methyl]phenyl}boronic acid
Synonyms: 2,6-bis[(2,2,6,6-tetramethylpiperidin-1-yl)methyl]phenylboronic acid
Molecular Weight (g/mol): 428.47
Molecular Formula: C26H45BN2O2
InChi Key: VSCRLRJQVRYOGB-UHFFFAOYSA-N
SMILES: CC1(C)CCCC(C)(C)N1CC1=CC=CC(CN2C(C)(C)CCCC2(C)C)=C1B(O)O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results