Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis[(2,2,6,6-tetramethyl-1-piperidinyl)methyl]phenylboronic Acid 97.0+%, TCI America™
SDP

Catalog No. B3927200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3927200MG 200 mg
B39271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3927200MG Supplier TCI America Supplier No. B3927200MG

(contains varying amounts of Anhydride)

Chemical Identifiers

CAS 1243264-54-1
Molecular Formula C26H45BN2O2
Molecular Weight (g/mol) 428.468
InChI Key VSCRLRJQVRYOGB-UHFFFAOYSA-N
PubChem CID 52912415
IUPAC Name [2,6-bis[(2,2,6,6-tetramethylpiperidin-1-yl)methyl]phenyl]boronic acid
SMILES B(C1=C(C=CC=C1CN2C(CCCC2(C)C)(C)C)CN3C(CCCC3(C)C)(C)C)(O)O

Specifications

Color White
Quantity 200 mg
Formula Weight 428.47
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis[(2,2,6,6-tetramethyl-1-piperidinyl)methyl]phenylboronic Acid (contains varying amounts of An
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.