CAS RN 1246659-36-8
CAS RN 1246659-36-8
IUPAC Name:
(2S)-3-(adamantan-1-yl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms:
(2S)-3-(adamantan-1-yl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol):
445.56
Molecular Formula:
C28H31NO4
InChi Key:
KKIZUCSOMHYCNI-SNXCOZLRSA-N
SMILES:
[H]C12CC3([H])CC([H])(C1)CC(C[C@H](NC(=O)OCC1C4=C(C=CC=C4)C4=C1C=CC=C4)C(O)=O)(C2)C3
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Ala Adamantyl -OH, Advanced ChemTech
CAS: 1246659-36-8 Molecular Formula: C28H31NO4 Molecular Weight (g/mol): 445.56 InChI Key: KKIZUCSOMHYCNI-SNXCOZLRSA-N IUPAC Name: (2S)-3-(adamantan-1-yl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid SMILES: [H]C12CC3([H])CC([H])(C1)CC(C[C@H](NC(=O)OCC1C4=C(C=CC=C4)C4=C1C=CC=C4)C(O)=O)(C2)C3