accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124750-51-2

IUPAC Name: 5-[4'-(bromomethyl)-[1,1'-biphenyl]-2-yl]-1-(triphenylmethyl)-1H-1,2,3,4-tetrazole
Synonyms: 5-[4'-(bromomethyl)-[1,1'-biphenyl]-2-yl]-1-(triphenylmethyl)-1,2,3,4-tetrazole
Molecular Weight (g/mol): 557.5
Molecular Formula: C33H25BrN4
InChi Key: ZTFVTXDWDFIQEU-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=C1)C1=CC=CC=C1C1=NN=NN1C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results