Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-1-triphenylmethyl-1H-tetrazole, TCI America™
SDP

Catalog No. B44625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44625G 5 g
B44621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44625G Supplier TCI America Supplier No. B44625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124750-51-2
Molecular Formula C33H25BrN4
Molecular Weight (g/mol) 557.495
MDL Number MFCD00665221
InChI Key ZTFVTXDWDFIQEU-UHFFFAOYSA-N
PubChem CID 10053461
IUPAC Name 5-[2-[4-(bromomethyl)phenyl]phenyl]-1-trityltetrazole
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C(=NN=N4)C5=CC=CC=C5C6=CC=C(C=C6)CBr

Specifications

Melting Point 147°C
Color White
Boiling Point 61°C
Quantity 5 g
UN Number 2811
Synonym 5-[4′-(Bromomethyl)-1,1′-biphenyl-2-yl]-1-trityl-1H-tetrazole
Formula Weight 557.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-[4′-(Bromomethyl)-1,1′-biphenyl-2-yl]-1-triphenylmethyl-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.