accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(R)(S)NHOO

CAS RN 124753-65-7

IUPAC Name: (1R,6S)-6-{[(benzyloxy)carbonyl]amino}cyclohex-3-ene-1-carboxylate
Synonyms: (1R,6S)-6-{[(benzyloxy)carbonyl]amino}cyclohex-3-ene-1-carboxylate
Molecular Weight (g/mol): 274.3
Molecular Formula: C15H16NO4
InChi Key: QIAAZWPFGHTXNZ-OLZOCXBDSA-M
SMILES: [O-]C(=O)[C@@H]1CC=CC[C@@H]1NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)