CAS RN 1247879-16-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris™ CycLuc 1
Synthetic luciferase substrate; brain penetrant; displays NIR emission