accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNNNOO(S)CH3H2NOOCH3NH2HCl

CAS RN 124832-27-5

IUPAC Name: 2-[(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate hydrochloride
Synonyms: valacyclovir hydrochloride
Molecular Weight (g/mol): 360.8
Molecular Formula: C13H21ClN6O4
InChi Key: ZCDDBUOENGJMLV-VGVXOIKUNA-N
SMILES: Cl.CC(C)[C@H](N)C(=O)OCCOCN1C=NC2=C1N=C(N)NC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)