accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124953-60-2

NOOCl

CAS RN 124953-60-2

IUPAC Name: 3-phenyl-1,2-oxazole-5-carbonyl chloride
Synonyms: 3-phenyl-1,2-oxazole-5-carbonyl chloride
Molecular Weight (g/mol): 207.61
Molecular Formula: C10H6ClNO2
InChi Key: HGXKTMWKFBNMMH-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results