accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNONCl

CAS RN 1249554-79-7

IUPAC Name: 5-(chloromethyl)-3-[(thiophen-2-yl)methyl]-1,2,4-oxadiazole
Synonyms: 5-(chloromethyl)-3-(thiophen-2-ylmethyl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 214.67
Molecular Formula: C8H7ClN2OS
InChi Key: GTOAVBZDTOHAGL-UHFFFAOYSA-N
SMILES: ClCC1=NC(CC2=CC=CS2)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results