accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1251-85-0

IUPAC Name: 4-[(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)methyl]-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Synonyms: 4-[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)methyl]-1,5-dimethyl-2-phenylpyrazol-3-one
Molecular Weight (g/mol): 388.47
Molecular Formula: C23H24N4O2
InChi Key: QATHNKNPUVVKHK-UHFFFAOYSA-N
SMILES: CN1N(C(=O)C(CC2=C(C)N(C)N(C2=O)C2=CC=CC=C2)=C1C)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)