Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diantipyrylmethane 99%, Thermo Scientific™

Catalog No. AC406420250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC406420250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC406420250 Supplier Thermo Scientific Chemicals Supplier No. 406420250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1251-85-0
Molecular Formula C23H24N4O2
Molecular Weight (g/mol) 388.471
InChI Key QATHNKNPUVVKHK-UHFFFAOYSA-N
Synonym diantipyrylmethane, 4,4'-methylenediantipyrine, bisantipyrylmethane, diantipyrinylmethane, 4,4'-methylenebis 1,5-dimethyl-2-phenyl-1h-pyrazol-3 2h-one, 4,4'-diantipyrylmethane, antipyrine, 4,4'-methylenedi, 3h-pyrazol-3-one, 4,4'-methylenebis 1,2-dihydro-1,5-dimethyl-2-phenyl, 4-1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1h-pyrazol-4-yl methyl-1,5-dimethyl-2-phenyl-2,3-dihydro-1h-pyrazol-3-one, 4-1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl methyl-1,5-dimethyl-2-phenylpyrazol-3-one
PubChem CID 96229
IUPAC Name 4-[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)methyl]-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)CC3=C(N(N(C3=O)C4=CC=CC=C4)C)C

Specifications

Melting Point 178°C to 180°C
Color White
Infrared Spectrum Authentic
Quantity 25 g
Beilstein 26, 489
Formula Weight 388.46
Percent Purity 98.5 to 101.5% (on dry substance)
Physical Form Crystalline Powder
Chemical Name or Material Diantipyrylmethane, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.