CAS RN 125110-14-7
IUPAC Name:
(6R,7R)-7-[(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-[(carboxymethoxy)imino]acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid trihydrate
Synonyms:
cefixim trihydrate
Molecular Weight (g/mol):
507.49
Molecular Formula:
C16H21N5O10S2
InChi Key:
IPYWNMVPZOAFOQ-XZVQIPOZSA-N
SMILES:
O.O.O.NC1=NC(=CS1)C(=N/OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O
1
–
1
of
1
results
1
Cefixime trihydrate
CAS: 125110-14-7 Molecular Formula: C16H15N5O7S2 Molecular Weight (g/mol): 453.44 MDL Number: MFCD00865020 InChI Key: OKBVVJOGVLARMR-QSWIMTSFSA-N PubChem CID: 5362065 ChEBI: CHEBI:472657 IUPAC Name: (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(carboxymethoxy)imino]acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid SMILES: NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O