Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefixime trihydrate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012840 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2840 25 mg
11-101-2839 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2840 Supplier MilliporeSigma Supelco Supplier No. 1858825MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical structure: ß-lactam

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD03788802

Specifications

CAS 125110-14-7
Linear Formula C16H15N5O7S2 · 3H2 O
Quantity 25 mg
Molecular Weight (g/mol) 507.50
Formula Weight 507.50
Percent Purity ≥98.0% (HPLC)
Grade For microbiology
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.