CAS RN 1251384-75-4
IUPAC Name:
(3-amino-2-methylpropyl)(methyl)amine
Synonyms:
(3-amino-2-methylpropyl)(methyl)amine
Molecular Weight (g/mol):
102.18
Molecular Formula:
C5H14N2
InChi Key:
HKAXXLXZPQLQHR-UHFFFAOYNA-N
SMILES:
CNCC(C)CN
1
–
1
of
1
results
1
N,2-Dimethyl-1,3-propanediamine 97.0+%, TCI America™
CAS: 1251384-75-4 Molecular Formula: C5H14N2 Molecular Weight (g/mol): 102.18 MDL Number: MFCD10566893 InChI Key: HKAXXLXZPQLQHR-UHFFFAOYNA-N PubChem CID: 19032309 IUPAC Name: (3-amino-2-methylpropyl)(methyl)amine SMILES: CNCC(C)CN