Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,2-Dimethyl-1,3-propanediamine 97.0+%, TCI America™
SDP

Catalog No. D36175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36175G Supplier TCI America Supplier No. D36175G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1251384-75-4
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD10566893
InChI Key HKAXXLXZPQLQHR-UHFFFAOYNA-N
PubChem CID 19032309
IUPAC Name (3-amino-2-methylpropyl)(methyl)amine
SMILES CNCC(C)CN

Specifications

Color Yellow
Quantity 5 g
UN Number 2734
Formula Weight 102.18
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,2-Dimethyl-1,3-propanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.