CAS RN 125229-61-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Alprazolam-d5 Solution, 100 μg/mL in methanol, ampule of 1 mL, Sold by MilliporeSigma™ Supelco™
An internal standard suitable for alprazolam testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, pain prescription monitoring or forensic analysis.