accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1256345-67-1

IUPAC Name: (4-cyano-2-methoxyphenyl)boronic acid
Synonyms: 4-cyano-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 176.97
Molecular Formula: C8H8BNO3
InChi Key: CCUBWDNQHCIXNF-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(=C1)C#N)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results