CAS RN 1256355-23-3
IUPAC Name:
[3-(dimethylamino)phenyl]boronic acid hydrochloride
Synonyms:
3-(dimethylamino)phenylboronic acid hydrochloride
Molecular Weight (g/mol):
201.46
Molecular Formula:
C8H13BClNO2
InChi Key:
QDYZZIRCCVRLBI-UHFFFAOYSA-N
SMILES:
Cl.CN(C)C1=CC=CC(=C1)B(O)O