Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D40135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4013-5G 5 g
D40131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4013-5G Supplier TCI America Supplier No. D40135G
Add to Cart
Add to Cart

Specifications

CAS 1256355-23-3
Color White
Molecular Formula C8H13BClNO2
MDL Number MFCD04112542
Quantity 5 g
Synonym 3-(Dimethylamino)benzeneboronic Acid Hydrochloride
InChI Key QDYZZIRCCVRLBI-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)N(C)C)(O)O.Cl
IUPAC Name [3-(dimethylamino)phenyl]boronic acid;hydrochloride
Molecular Weight (g/mol) 201.457
PubChem CID 44754897
Formula Weight 201.46
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.