accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNH2FHOOHHCl

CAS RN 1256355-32-4

IUPAC Name: (4-amino-3-fluorophenyl)boronic acid hydrochloride
Synonyms: 4-amino-3-fluorophenylboronic acid hydrochloride
Molecular Weight (g/mol): 191.39
Molecular Formula: C6H8BClFNO2
InChi Key: NXCNVCKJELIVDM-UHFFFAOYSA-N
SMILES: Cl.NC1=C(F)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results