accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1256360-51-6

IUPAC Name: 4-{[2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}morpholine
Synonyms: 4-{[2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}morpholine
Molecular Weight (g/mol): 321.2
Molecular Formula: C17H25BFNO3
InChi Key: VSRFPDFCDOCPEQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C(CN2CCOCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results