accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 125697-92-9

HOOOHNHOOHOH

CAS RN 125697-92-9

IUPAC Name: 5-{[(2,5-dihydroxyphenyl)methyl][(2-hydroxyphenyl)methyl]amino}-2-hydroxybenzoic acid
Synonyms: 5-{[(2,5-dihydroxyphenyl)methyl][(2-hydroxyphenyl)methyl]amino}-2-hydroxybenzoic acid
Molecular Weight (g/mol): 381.38
Molecular Formula: C21H19NO6
InChi Key: ULTTYPMRMMDONC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1O)N(CC1=CC=CC=C1O)CC1=CC(O)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results