Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lavendustin A, R&D Systems™
SDP

Catalog No. 13311 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-311 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-311 Supplier R&D Systems Supplier No. LG5931102

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. EGFR, p60c-src inhibitor

Potent, cell-permeable inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase (IC50 = 11 nM). Inhibits p60c-src with an IC50 of 500 nM and is selective over PKA, PKC and PI 3-kinase (IC50 > 100 μM).

Chemical Identifiers

CAS 125697-92-9
Molecular Formula C21H19NO6
Molecular Weight (g/mol) 381.384
InChI Key ULTTYPMRMMDONC-UHFFFAOYSA-N
Synonym lavendustin a, unii-3y0g32g2rv, lavendustin a, from streptomyces griseolavendus, benzoic acid,5-2,5-dihydroxyphenyl methyl 2-hydroxyphenyl methyl amino-2-hydroxy, 5-2,5-dihydroxyphenyl methyl 2-hydroxyphenyl methyl amino-2-hydroxybenzoic acid, benzoic acid, 5-2,5-dihydroxyphenyl methyl 2-hydroxyphenyl methyl amino-2-hydroxy, 5-2,5-dihydroxybenzyl 2-hydroxybenzyl amino-2-hydroxybenzoic acid, 5-amino-n-2,5-dihydroxybenzyl-n?-2-hydroxybenzyl salicylic acid, 5-amino-n-2,5-dihydroxybenzyl-n′-2-hydroxybenzyl salicylic acid
PubChem CID 3894
IUPAC Name 5-[(2,5-dihydroxyphenyl)methyl-[(2-hydroxyphenyl)methyl]amino]-2-hydroxybenzoic acid
SMILES C1=CC=C(C(=C1)CN(CC2=C(C=CC(=C2)O)O)C3=CC(=C(C=C3)O)C(=O)O)O

Specifications

Quantity 1 mg
Formula Weight 381.38
Percent Purity >98%
Product Line Tocris
Chemical Name or Material Lavendustin A
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.