accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOH(R)OOOHOHOH(R)OOHOH

CAS RN 1257-08-5

IUPAC Name: (2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-1-benzopyran-3-yl 3,4,5-trihydroxybenzoate
Synonyms: epicatechin gallate (-)-epicatechin 3-O-gallate
Molecular Weight (g/mol): 442.38
Molecular Formula: C22H18O10
InChi Key: LSHVYAFMTMFKBA-TZIWHRDSSA-N
SMILES: OC1=CC(O)=C2C[C@@H](OC(=O)C3=CC(O)=C(O)C(O)=C3)[C@H](OC2=C1)C1=CC=C(O)C(O)=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results