CAS RN 125700-67-6
IUPAC Name:
[(1H-1,2,3-benzotriazol-1-yloxy)(dimethylamino)methylidene]dimethylazanium; tetrafluoroboranuide
Synonyms:
[(1,2,3-benzotriazol-1-yloxy)(dimethylamino)methylidene]dimethylazanium tetrafluoroborate
Molecular Weight (g/mol):
321.09
Molecular Formula:
C11H16BF4N5O
InChi Key:
JKEKMBGUVUKMQB-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C12)=[N+](C)C
1
–
4
of
4
results
1
Promotions Available
O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99%