CAS RN 1258003-96-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Granzyme B Inhibitor II
The Granzyme B Inhibitor II, also referenced under CAS 1258003-96-1, controls the biological activity of Granzyme B. This small molecule/inhibitor is primarily used for Cancer applications.