Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Granzyme B Inhibitor II

Catalog No. 3680551MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-805-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-805-51MG Supplier MilliporeSigma™ Supplier No. 3680551MG
Add to Cart
Add to Cart

The Granzyme B Inhibitor II, also referenced under CAS 1258003-96-1, controls the biological activity of Granzyme B. This small molecule/inhibitor is primarily used for Cancer applications.

Peptide Sequence: Ac-Ile-Glu-Thr-Asp-CHO
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins caspase-8
CAS 1258003-96-1
Molecular Formula C21H34N4O10
Formula Weight 502.5g/mol
Solubility DMSO
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.