CAS RN 1259484-97-3
IUPAC Name:
N'-[1-(3,4-dimethoxyphenyl)ethylidene]-3-(4-methoxyphenyl)-1H-pyrazole-5-carbohydrazide
Synonyms:
N'-[1-(3,4-dimethoxyphenyl)ethylidene]-5-(4-methoxyphenyl)-2H-pyrazole-3-carbohydrazide
Molecular Weight (g/mol):
394.43
Molecular Formula:
C21H22N4O4
InChi Key:
GMFUWEBOUKIKRP-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=NNC(=C1)C(=O)NN=C(C)C1=CC(OC)=C(OC)C=C1
1
–
1
of
1
results
1
SKI 178, Tocris Bioscience™
Chemical Name or Material: N'-(1-(3,4-Dimethoxyphenyl)ethylidene)-3-(4-methoxyphenyl)-1H-pyrazole-5-carbohydrazide Physical Form: Solid Synonym: SKI 178 Recommended Storage: Store at −20°C Solubility Information: Soluble to 100mM in DMSO Formula Weight: 394.42 Linear Formula: C21H22N4O4 CAS: 1259484-97-3