accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126-58-9

IUPAC Name: 2-{[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl}-2-(hydroxymethyl)propane-1,3-diol
Synonyms: dipentaerythritol
Molecular Weight (g/mol): 254.28
Molecular Formula: C10H22O7
InChi Key: TXBCBTDQIULDIA-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)COCC(CO)(CO)CO
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)