accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126-68-1

IUPAC Name: O,O,O-triethyl phosphorothioate
Synonyms: O,O,O-triethyl phosphorothioate
Molecular Weight (g/mol): 198.22
Molecular Formula: C6H15O3PS
InChi Key: QTPJMTACJMLPLL-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results