accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126-72-7

IUPAC Name: tris(2,3-dibromopropyl) phosphate
Synonyms: tris(2,3-dibromopropyl) phosphate
Molecular Weight (g/mol): 697.61
Molecular Formula: C9H15Br6O4P
InChi Key: PQYJRMFWJJONBO-UHFFFAOYNA-N
SMILES: BrCC(Br)COP(=O)(OCC(Br)CBr)OCC(Br)CBr
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results