Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris-(2,3-dibromopropyl) Phosphate, MP Biomedicals

Catalog No. ICN219625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219625 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ICN219625 Supplier MP Biomedicals, Inc Supplier No. 05219625.1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 126-72-7
Molecular Formula C9H15Br6O4P
Molecular Weight (g/mol) 697.613
InChI Key PQYJRMFWJJONBO-UHFFFAOYSA-N
Synonym tris 2,3-dibromopropyl phosphate, tdbpp, anfram 3pb, flacavon r, flammex ap, zetofex zn, tris flame retardant, tris-bp, tdbp
PubChem CID 31356
ChEBI CHEBI:82282
IUPAC Name tris(2,3-dibromopropyl) phosphate
SMILES C(C(CBr)Br)OP(=O)(OCC(CBr)Br)OCC(CBr)Br

Specifications

Quantity 100 g
Formula Weight 697.64
Chemical Name or Material Tris-(2, 3-Dibromopropyl)Phosphate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.