accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COO

CAS RN 126-81-8

IUPAC Name: 5,5-dimethylcyclohexane-1,3-dione
Synonyms: medon dimedone
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: BADXJIPKFRBFOT-UHFFFAOYSA-N
SMILES: CC1(C)CC(=O)CC(=O)C1
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
Quantity 
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (4)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (3)
  • (7)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)