Learn More
5,5-Dimethyl-1,3-cycloHexanedione, For HPLC derivatization, ≥99.0%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 126-81-8 |
| Melting Point | 146°C to 148°C |
| Linear Formula | (CH3)2C6H6(=O)2 |
| Quantity | 50 g |
| Molecular Weight (g/mol) | 140.18 |
| Formula Weight | 140.18 |
| Percent Purity | ≥99.0% (GC); ≥99.0% |
| Grade | For HPLC derivatization |
| Physical Form | Solid |
Safety and Handling
| RTECSNumber | GV0345000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.