accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3CH3OH3C

CAS RN 126-84-1

IUPAC Name: 2,2-diethoxypropane
Synonyms: 2,2-diethoxypropane
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: FGQLGYBGTRHODR-UHFFFAOYSA-N
SMILES: CCOC(C)(C)OCC
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)