Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diethoxypropane, Technical Grade, Thermo Scientific™

Catalog No. AAB2403522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2403522 100 g
AAB2403506 5 g
AAB2403514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2403522 Supplier Thermo Scientific Chemicals Supplier No. B2403522

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-84-1
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.203
MDL Number MFCD00009224
InChI Key FGQLGYBGTRHODR-UHFFFAOYSA-N
Synonym acetone diethyl acetal, propane, 2,2-diethoxy, acetone diethyl ketal, acetone, diethyl acetal, usaf do-44, unii-18i1qhh2k2, acetonediethylketal, acetone diethylacetal, diethoxypropane, propane,2-diethoxy
PubChem CID 31361
IUPAC Name 2,2-diethoxypropane
SMILES CCOC(C)(C)OCC

Specifications

Boiling Point 44°C to 46°C (60 mmHg)
Flash Point 7°C (45°F)
Quantity 100 g
UN Number UN3271
Formula Weight 132.2
Grade Technical
Chemical Name or Material 2,2-Diethoxypropane
Hazard Category Danger
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.