accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CONNFFFONHOHNOH

CAS RN 1260181-14-3

IUPAC Name: 3-{5-[4-(2-hydroxy-2-methylpropanoyl)piperazin-1-yl]-2-(trifluoromethyl)phenyl}-4-(1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 3-{5-[4-(2-hydroxy-2-methylpropanoyl)piperazin-1-yl]-2-(trifluoromethyl)phenyl}-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione
Molecular Weight (g/mol): 526.52
Molecular Formula: C27H25F3N4O4
InChi Key: CLGRAWDGLMENOD-UHFFFAOYSA-N
SMILES: CC(C)(O)C(=O)N1CCN(CC1)C1=CC(C2=C(C(=O)NC2=O)C2=CNC3=CC=CC=C23)=C(C=C1)C(F)(F)F
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results