accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNBr(E/Z)

CAS RN 126115-86-4

IUPAC Name: 1-octadecyl-4-(2-phenylethenyl)pyridin-1-ium bromide
Synonyms: 1-octadecyl-4-(2-phenylethenyl)pyridin-1-ium bromide
Molecular Weight (g/mol): 514.64
Molecular Formula: C31H48BrN
InChi Key: FEIYCOMNTWABLV-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCCCC[N+]1=CC=C(C=CC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results