Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Octadecyl-4-stilbazole Bromide 97.0+%, TCI America™
SDP

Catalog No. O01921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01921G Supplier TCI America Supplier No. O01921G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 126115-86-4
Molecular Formula C31H48BrN
Molecular Weight (g/mol) 514.636
MDL Number MFCD00060044
InChI Key FEIYCOMNTWABLV-UHFFFAOYSA-M
Synonym N-Stearyl-4-stilbazole Bromide
PubChem CID 70700167
IUPAC Name 1-octadecyl-4-(2-phenylethenyl)pyridin-1-ium;bromide
SMILES CCCCCCCCCCCCCCCCCC[N+]1=CC=C(C=C1)C=CC2=CC=CC=C2.[Br-]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 514.64
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Octadecyl-4-stilbazole Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.