accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH3CPH3CCH3H3CCH3H3CCH3CH3CH3CH3CH3CH3H3C(?)

CAS RN 1262046-34-3

IUPAC Name: di-tert-butyl[3-methoxy-6-methyl-2',4',6'-tris(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Synonyms: di-tert-butyl({2',4',6'-triisopropyl-3-methoxy-6-methyl-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 468.71
Molecular Formula: C31H49OP
InChi Key: CVLLAKCGAFNZHJ-UHFFFAOYSA-N
SMILES: COC1=C(P(C(C)(C)C)C(C)(C)C)C(=C(C)C=C1)C1=C(C=C(C=C1C(C)C)C(C)C)C(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results