Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(2,2,2-Trifluoroethyl) Borate

Catalog No. 7908771G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-087-71G 2 g
79-087-710G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 79-087-71G Supplier Sigma Aldrich Supplier No. 7908771G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent can promote the direct formation of amides from carboxylic acids and amines under thermal and microwave conditions.

Specifications

CAS 1262046-34-3
Melting Point 121°C to 131°C
Molecular Formula C31H49OP
Linear Formula C31H49OP
Refractive Index n/D 1.298
MDL Number MFCD20922895
Quantity 2 g
Synonym Tris(2,2,2-trifluoroethyl)borate; Sheppard Amidation Reagent; Tris(2,2,2-trifluoroethoxy)boron; Sheppard Reagent
Molecular Weight (g/mol) 468.69 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.