accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126213-50-1

IUPAC Name: 2H,3H-thieno[3,4-b][1,4]dioxine
Synonyms: 2H,3H-thieno[3,4-b][1,4]dioxine
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: GKWLILHTTGWKLQ-UHFFFAOYSA-N
SMILES: C1COC2=CSC=C2O1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)