Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4-Ethylenedioxythiophene

Catalog No. 48302810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-302-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-302-810G Supplier Sigma Aldrich Supplier No. 48302810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Monomer used in the synthesis of conducting polymers.EDOT can be polymerized to form poly(3,4-ethylenedioxythiophene) (PEDOT) for use as an electrochromic polymer (EC) based coating for a variety of applications like solid state organic electrochemical supercapacitors (OESCs), electrochromic devices (ECDs), and carbon nanotubes (CNTs) based electrochemical devices for diabetes monitoring. It can be used:As a reductant in a one-pot synthesis of gold nanoparticles from HAuCl4 (254169).As a starting material used in palladium-catalyzed mono- and bis-arylation reactions.In the synthesis of conjugated polymers and copolymers, with potential optical applications.

Specifications

CAS 126213-50-1
Density 1.331 g/mL (at 25°C)
Boiling Point 193°C
Molecular Formula C6H6O2S
Refractive Index n20/D 1.5765 (literature)
Linear Formula C6H6O2S
Quantity 10 g
MDL Number MFCD02093622
Synonym 2,3-Dihydrothieno[3,4-b]-1,4-dioxin; EDOT
Molecular Weight (g/mol) 142.18
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.