accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CH3CH3CCNNN

CAS RN 1262230-03-4

IUPAC Name: [(cyanoimino)methylidene]azanide; tributyl(methyl)azanium
Synonyms: [(cyanoimino)methylidene]azanide; tributyl(methyl)azanium
Molecular Weight (g/mol): 266.43
Molecular Formula: C15H30N4
InChi Key: FMYCKRQZPVZFLW-UHFFFAOYSA-N
SMILES: [N-]=C=NC#N.CCCC[N+](C)(CCCC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results