Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl(methyl)ammonium Dicyanamide 98.0+%, TCI America™
SDP

Catalog No. T35335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T35335G Supplier TCI America Supplier No. T35335G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1262230-03-4
Molecular Formula C15H30N4
Molecular Weight (g/mol) 266.433
InChI Key FMYCKRQZPVZFLW-UHFFFAOYSA-N
PubChem CID 121234265
IUPAC Name cyanoiminomethylideneazanide;tributyl(methyl)azanium
SMILES CCCC[N+](C)(CCCC)CCCC.C(=[N-])=NC#N

Specifications

Melting Point 12°C
Color Yellow
Quantity 5 g
Formula Weight 266.43
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material Tributyl(methyl)ammonium Dicyanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.