accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrFFFF

CAS RN 1262412-30-5

IUPAC Name: 5-bromo-3-fluoro-2-(trifluoromethyl)pyridine
Synonyms: 5-bromo-3-fluoro-2-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 243.99
Molecular Formula: C6H2BrF4N
InChi Key: ZYPJKTSNQGVERF-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CN=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results